Thermo Scientific Chemicals

2-Phenyl-2-propanol, 98+%, Thermo Scientific Chemicals

Catalog number: A15580.22
100 g, Each
Thermo Scientific Chemicals

2-Phenyl-2-propanol, 98+%, Thermo Scientific Chemicals

Catalog number: A15580.22
100 g, Each
Quantity
Catalog number: A15580.22
also known as A15580-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-94-7
IUPAC Name
2-phenylpropan-2-ol
Molecular Formula
C9H12O
InChI Key
BDCFWIDZNLCTMF-UHFFFAOYSA-N
SMILES
CC(C)(O)C1=CC=CC=C1
Appearance (Color)
Colorless to white to pale yellow
Form
Crystals or powder or crystalline powder or clear liquid as melt
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
26.0-35.0°C

Description

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.

Solubility
Insoluble in water.

Notes
Store away from acids, acid chlorides, acid anhydrides, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text