Thermo Scientific Chemicals

p-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: A15581.22
100 g, Each
Thermo Scientific Chemicals

p-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: A15581.22
100 g, Each
Quantity
Catalog number: A15581.22
also known as A15581-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
874-60-2
IUPAC Name
4-methylbenzoyl chloride
Molecular Formula
C8H7ClO
InChI Key
NQUVCRCCRXRJCK-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.5525-1.5555 @ 20?C

Description

p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

Solubility
Reacts with water alcohol.

Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text