p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.
Solubility
Reacts with water alcohol.
Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
RUO – Research Use Only
M H Karol; Y Alarie. Kaputsky. Antigens which detect IgE antibodies in workers sensitive to toluene diisocyanate. Clinical allergy. 1980,10 (1), 101-109.
Shainaz M Landge; M Chidambaram; A.P Singh. Benzoylation of toluene with p-toluoyl chloride over triflic acid functionalized mesoporous Zr-TMS catalyst. Journal of Molecular Catalysis A: Chemical 2004, 213 (2), 257-266.