Sucrose, 99%
Thermo Scientific Chemicals

Sucrose, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
10,000 g
Catalog number A15583.0E
also known as A15583-0E
Price (USD)/ Each
83.65
Online exclusive
93.20 
Save 9.55 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
83.65
Online exclusive
93.20 
Save 9.55 (10%)
Add to cart
Sucrose, 99%
Catalog numberA15583.0E
Price (USD)/ Each
83.65
Online exclusive
93.20 
Save 9.55 (10%)
-
Add to cart
Chemical Identifiers
CAS57-50-1
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyCZMRCDWAGMRECN-PWPRYFECNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (unspecified)≥98.5%
Identification (FTIR)Conforms (non-U.S. sourced material)
Optical Rotation+66.5° ± 1° (c=10 in water, non-U.S. sourced material)
FormCrystals or crystalline powder
View more
Used for protein purification in sucrose density gradient studiesSucrose is widely utilized in medications to carry a more pleasant taste to indigestible chemicals. It is used in many medical dosage forms such as chewable tablets, syrups, tablets, or glues. Sucrose is mainly used in foods nowadays. The fatty acid esters of sucrose show useful application in transdermal therapeutic systems. Sucrose-gradient centrifugation also plays an important role in the selective isolation of nocardia spp. from soil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in alcohol, methanol. Insoluble in diethyl ether.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Jain, A.; Shore, A. M.; Jonnalagadda, S. C.; Ramanujachary, K. V.; Mugweru, A. Conversion of fructose, glucose and sucrose to 5-hydroxymethyl-2-furfural over mesoporous zirconium phosphate catalyst. Appl. Catal. A 2015, 489, 72-76.
  2. Chen, Y.; Zhang, H.; Li, A.; Zhuo, K. Conductivities for ZnSO4/MgSO4+ sucrose/trehalose + water systems at 298.15 K. Fluid Phase Equilib. 2015, 388, 78-83.