Gibco Primary Mouse Cortical Neurons are isolated and cryopreserved from C57BL/6 embryonic day-17 mice. A flexible, ready-to-use, and quality alternativeRead more
Have Questions?
Change view
Catalog Number
Quantity
A15586
4 x 106
A15585
1 x 106
Catalog number A15586
Price (USD)/ 4 x 106 cells
742.65
Online exclusive
800.00
Save 57.35 (7%)
-
Add to cart
Quantity:
4 x 106
Price (USD)/ 4 x 106 cells
742.65
Online exclusive
800.00
Save 57.35 (7%)
Add to cart
Primary Mouse Cortical Neurons
Catalog numberA15586
Price (USD)/ 4 x 106 cells
742.65
Online exclusive
800.00
Save 57.35 (7%)
-
Add to cart
Gibco Primary Mouse Cortical Neurons are isolated and cryopreserved from C57BL/6 embryonic day-17 mice. A flexible, ready-to-use, and quality alternative to fresh neurons, each vial contains 1 million cells per mL. These neurons consistently yield post-thaw viability in the range of 50-80%, have high neuronal purity, and are validated for functionality.
• High viability: post-thaw viability of 50% or greater • High purity: greater than 98% neuronal phenotype • Functionally validated: tested for long-term survival, neurotransmitter response, and transduction
For Research Use Only. Not for use in diagnostic procedures.