Thermo Scientific Chemicals

Ethyl 2,3-dibromopropionate, 97%, Thermo Scientific Chemicals

Catalog number: A15591.14
25 g, Each
Thermo Scientific Chemicals

Ethyl 2,3-dibromopropionate, 97%, Thermo Scientific Chemicals

Catalog number: A15591.14
25 g, Each
Quantity
Catalog number: A15591.14
also known as A15591-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3674-13-3
IUPAC Name
ethyl 2,3-dibromopropanoate
Molecular Formula
C5H8Br2O2
InChI Key
OENICUBCLXKLJQ-UHFFFAOYNA-N
SMILES
CCOC(=O)C(Br)CBr
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.4985-1.5035 @ 20?C

Description

Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text