Thermo Scientific Chemicals

4-Picoline, 98%, Thermo Scientific Chemicals

Catalog number: A15595.AE
100 mL, Each
Thermo Scientific Chemicals

4-Picoline, 98%, Thermo Scientific Chemicals

Catalog number: A15595.AE
100 mL, Each
Quantity
Catalog number: A15595.AE
also known as A15595-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-89-4
IUPAC Name
4-methylpyridine
Molecular Formula
C6H7N
InChI Key
FKNQCJSGGFJEIZ-UHFFFAOYSA-N
SMILES
CC1=CC=NC=C1
Appearance (Color)
Clear colorless to yellow
Assay from Supplier's CofA
≥97.5% (US Sourced Material)
Identification (FTIR)
Conforms (UK Sourced Material)
Comment
Material Sourced in UK and US
Assay (GC)
≥97.5% (UK Sourced Material)

Description

4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

Solubility
Soluble in water, alcohol and ether.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text