Ethyl oleate, tech. 70%, Thermo Scientific Chemicals
Ethyl oleate, tech. 70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl oleate, tech. 70%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
5000 g
Catalog number A15601.0B
also known as A15601-0B
Price (USD)/ Each
103.00
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
103.00
Add to cart
Ethyl oleate, tech. 70%, Thermo Scientific Chemicals
Catalog numberA15601.0B
Price (USD)/ Each
103.00
-
Add to cart
Chemical Identifiers
CAS111-62-6
IUPAC Nameethyl (9E)-octadec-9-enoate
Molecular FormulaC20H38O2
InChI KeyLVGKNOAMLMIIKO-VAWYXSNFSA-N
SMILESCCCCCCCC\C=C\CCCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4470-1.4550 @ 20?C
Assay (GC)>68.0%
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only