Isopropylidene malonate, 97%, Thermo Scientific Chemicals
Isopropylidene malonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropylidene malonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A15603.22
also known as A15603-22
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
Add to cart
Isopropylidene malonate, 97%, Thermo Scientific Chemicals
Catalog numberA15603.22
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
-
Add to cart
Chemical Identifiers
CAS2033-24-1
IUPAC Name2,2-dimethyl-1,3-dioxane-4,6-dione
Molecular FormulaC6H8O4
InChI KeyGXHFUVWIGNLZSC-UHFFFAOYSA-N
SMILESCC1(C)OC(=O)CC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
Melting Point (clear melt)88-97°C
FormCrystals or crystalline powder
Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.

Solubility
Soluble in water. Slightly soluble in ethanol and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Malonate ester which is an unusually strong acid (pKa 9.97) in comparison with, e.g. Diethyl malonate, A15468 : J. Am. Chem. Soc., 70, 3426 (1948). The mild conditions required for alkylation, acylation, etc. and the ease with which the products undergo alcoholysis or hydrolysis-decarboxylation make Meldrum's acid a valuable and versatile synthetic intermediate. Reviews: Chem. Soc. Rev., 7, 345 (1978); Heterocycles, 32, 529 (1991); Synlett, 1649 (2004).
  2. For phase-transfer alkylation using K2CO3 as base, see: Synthesis, 452 (1982); Bull. Soc. Chim. Belg., 104, 643 (1995). Phase-transfer addition to Michael acceptors also occurs: Synthesis, 224 (1984). Mitsunobu dialkylation with an alcohol and DIAD/Ph3P is also possible: J. Org. Chem., 62, 1617 (1997).
  3. Knoevenagel condensation with aldehydes gives ylidene derivatives which are solvolyzed by alcohols to give monoesters of alkylidene or arylidene malonic acids: J. Am. Chem. Soc., 74, 5897 (1952). For diastereoselective condensation with citronellal, followed by intramolecular Diels-Alder reaction, see: Org. Synth. Coll., 8, 353 (1993). With formaldehyde in pyridine gives the stable pyridinium methylide which, on treatment with acid, generates the useful reactive Michael acceptor and dienophile, methylene Meldrum's acid: Synthesis, 215 (1996).