Thermo Scientific Chemicals

3'-Methoxyacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: A15604.22
100 g, Each
Thermo Scientific Chemicals

3'-Methoxyacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: A15604.22
100 g, Each
Quantity
Catalog number: A15604.22
also known as A15604-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
586-37-8
IUPAC Name
1-(3-methoxyphenyl)ethan-1-one
Molecular Formula
C9H10O2
InChI Key
BAYUSCHCCGXLAY-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)C(C)=O
Form
Liquid
Refractive Index
1.5380-1.5430 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%

Description

It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

Solubility
Fully miscible in water. Soluble in alcohol.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text