It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.
Solubility
Fully miscible in water. Soluble in alcohol.
Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Jorge A. Pino, et al. Aromatic Plants from Western Cuba VII. Composition of the Leaf Oils of Psidium wrightii Krug et Urb., Lantana involucrata L., Cinnamomum montanum (Sw.) Berchtold et J. Persl. And Caesalpinia violacea (Mill.) Standley.J. Essent. Oil Res.,2006,18(2), 170-174.
- Cesar A. Madrigal, et al. Asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes bearing a chiral phosphinoferrocenyloxazoline ligand.J. Organomet. Chem.,2008,693(15), 2535-2540.