Thermo Scientific Chemicals

Ethyl malonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A15616.06
5 g, Each
Thermo Scientific Chemicals

Ethyl malonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A15616.06
5 g, Each
Quantity
Catalog number: A15616.06
also known as A15616-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36239-09-5
IUPAC Name
ethyl 3-chloro-3-oxopropanoate
Molecular Formula
C5H7ClO3
InChI Key
KWFADUNOPOSMIJ-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(Cl)=O
Assay (Titration ex Chloride)
≥94.0 to ≤106.0%
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4265-1.4325 @ 20?C

Description

Ethyl malonyl chloride is used in the syntheses of methanofullerodendimers and 3-pyrrolin-2-ones. It plays a vital role in the preparation of 3,5-disubstituted 1,2,4-oxadiazole derivatives, which are potential peptidomimetic building blocks. It is a versatile acylating agent for propargyl alcohols, hydrazines and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl malonyl chloride is used in the syntheses of methanofullerodendimers and 3-pyrrolin-2-ones. It plays a vital role in the preparation of 3,5-disubstituted 1,2,4-oxadiazole derivatives, which are potential peptidomimetic building blocks. It is a versatile acylating agent for propargyl alcohols, hydrazines and amines.

Solubility
Miscible with water.

Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols. It reacts with water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text