Thermo Scientific Chemicals

4-Fluorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A15624.22
100 g, Each
Thermo Scientific Chemicals

4-Fluorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A15624.22
100 g, Each
Quantity
Catalog number: A15624.22
also known as A15624-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
823-85-8
IUPAC Name
hydrogen (4-fluorophenyl)hydrazine chloride
Molecular Formula
C6H8ClFN2
InChI Key
FEKUXLUOKFSMRO-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=C(F)C=C1
Appearance (Color)
White to cream to brown
Assay (Argentometric Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder

Description

4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.

Solubility
Soluble in water.

Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text