Thermo Scientific Chemicals

3-Bromoanisole, 98+%, Thermo Scientific Chemicals

Catalog number: A15625.22
100 g, Each
Thermo Scientific Chemicals

3-Bromoanisole, 98+%, Thermo Scientific Chemicals

Catalog number: A15625.22
100 g, Each
Quantity
Catalog number: A15625.22
also known as A15625-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2398-37-0
IUPAC Name
1-bromo-3-methoxybenzene
Molecular Formula
C7H7BrO
InChI Key
PLDWAJLZAAHOGG-UHFFFAOYSA-N
SMILES
COC1=CC=CC(Br)=C1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Form
Liquid
Refractive Index
1.5620-1.5660 @ 20?C

Description

3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

Solubility
Miscible in methanol, benzene, hexane, toluene and dichloromethane. Practically insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text