Thermo Scientific Chemicals

4-Chloropyridine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15628.14
25 g, Each
Thermo Scientific Chemicals

4-Chloropyridine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15628.14
25 g, Each
Quantity
Catalog number: A15628.14
also known as A15628-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7379-35-3
IUPAC Name
hydrogen 4-chloropyridine chloride
Molecular Formula
C5H5Cl2N
InChI Key
XGAFCCUNHIMIRV-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClC1=CC=NC=C1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to cream or pale yellow
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%

Description

It is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text