Thermo Scientific Chemicals

(2-Chloroethyl)trimethylammonium chloride, 98% (dry wt.), Thermo Scientific Chemicals

Catalog number: A15630.22
100 g, Each
Thermo Scientific Chemicals

(2-Chloroethyl)trimethylammonium chloride, 98% (dry wt.), Thermo Scientific Chemicals

Catalog number: A15630.22
100 g, Each
Quantity
Catalog number: A15630.22
also known as A15630-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
999-81-5
IUPAC Name
(2-chloroethyl)trimethylazanium chloride
Molecular Formula
C5H13Cl2N
InChI Key
UHZZMRAGKVHANO-UHFFFAOYSA-M
SMILES
[Cl-].C[N+](C)(C)CCCl
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (dry wt. basis) (UK-sourced Material)
Assay from Supplier's CofA
≥97.5% (US-sourced material)
Comment
Material sourced in UK and US

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text