Thermo Scientific Chemicals

4-Iodotoluene, 98%, Thermo Scientific Chemicals

Catalog number: A15631.14
25 g, Each
Thermo Scientific Chemicals

4-Iodotoluene, 98%, Thermo Scientific Chemicals

Catalog number: A15631.14
25 g, Each
Quantity
Catalog number: A15631.14
also known as A15631-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
624-31-7
IUPAC Name
1-iodo-4-methylbenzene
Molecular Formula
C7H7I
InChI Key
UDHAWRUAECEBHC-UHFFFAOYSA-N
SMILES
CC1=CC=C(I)C=C1
Melting Point (clear melt)
30-39?C
Appearance (Color)
White to yellow
Assay (GC)
≥97.5%
Form
Crystalline or fused solid
Identification (FTIR)
Conforms

Description

4-Iodotoluene is used in wide range of medicals industrial applications as well as in pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodotoluene is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals.

Solubility
Solubility in methanol is almost transparent.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text