Thermo Scientific Chemicals

2-Bromo-1,4-dimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog number: A15640.09
10 g, Each
Thermo Scientific Chemicals

2-Bromo-1,4-dimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog number: A15640.09
10 g, Each
Quantity
Catalog number: A15640.09
also known as A15640-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25245-34-5
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5685-1.5725 @ 20?C

Description

2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text