Thermo Scientific Chemicals

Methyltriphenylphosphonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: A15644.22
100 g, Each
Thermo Scientific Chemicals

Methyltriphenylphosphonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: A15644.22
100 g, Each
Quantity
Catalog number: A15644.22
also known as A15644-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2065-66-9
IUPAC Name
methyltriphenylphosphanium iodide
Molecular Formula
C19H18IP
InChI Key
JNMIXMFEVJHFNY-UHFFFAOYSA-M
SMILES
[I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Assay (Titration ex Iodide)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤1%
Appearance (Color)
White

Description

Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium iodide is as a reactant for synthesis of triphenylamine-based dyes and polyolefinic aromatic molecules with pyrene for use in dye-sensitized solar cells. Used as ligand in coupling reaction. It is also used as a precursor to a Wittig reagent.

Solubility
Soluble in water, acetone, dichloromethane , methanol.

Notes
Light sensitive and hygroscopic. Store in dark. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text