Thermo Scientific Chemicals

3,4-Dichlorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15653.14
25 g, Each
Thermo Scientific Chemicals

3,4-Dichlorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15653.14
25 g, Each
Quantity
Catalog number: A15653.14
also known as A15653-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5807-30-7
IUPAC Name
2-(3,4-dichlorophenyl)acetic acid
Molecular Formula
C8H6Cl2O2
InChI Key
ZOUPGSMSNQLUNW-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(Cl)C(Cl)=C1
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
84.0-91.0?C
Form
Crystals or powder or crystalline powder or lumps

Description

It is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text