Thermo Scientific Chemicals

Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A15656.36
500 g, Each
Thermo Scientific Chemicals

Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A15656.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15656.36
also known as A15656-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13755-38-9
IUPAC Name
disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Molecular Formula
C5H4FeN6Na2O3
InChI Key
JFPDVRGGFBUORC-UHFFFAOYSA-N
SMILES
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
10.7-14.7% (1.75 to 2.5 waters)
Identification (FTIR)
Conforms
Appearance (Color)
Brown to red to dark red
Assay (Argentometric Titration)
≥98.0% to ≤102.0% (as dihydrate)

Description

Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

Solubility
Soluble in water. Slightly soluble ethanol.

Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text