Thermo Scientific Chemicals

Bis(trimethylsilyl)methane, 98%, Thermo Scientific Chemicals

Catalog number: A15662.14
25 g, Each
Thermo Scientific Chemicals

Bis(trimethylsilyl)methane, 98%, Thermo Scientific Chemicals

Catalog number: A15662.14
25 g, Each
Quantity
Catalog number: A15662.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2117-28-4
IUPAC Name
trimethyl[(trimethylsilyl)methyl]silane
Molecular Formula
C7H20Si2
InChI Key
GYIODRUWWNNGPI-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C[Si](C)(C)C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Refractive Index
1.4150-1.4190 @ 20?C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text