Thermo Scientific Chemicals

4-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Catalog number: A15664.06
5 g, Each
Thermo Scientific Chemicals

4-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Catalog number: A15664.06
5 g, Each
Quantity
Catalog number: A15664.06
also known as A15664-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51135-96-7
IUPAC Name
4-benzyl-4-hydroxypiperidin-1-ium
Molecular Formula
C12H18NO
InChI Key
KJZBZOFESQSBCV-UHFFFAOYSA-O
SMILES
OC1(CC2=CC=CC=C2)CC[NH2+]CC1
Appearance (Color)
White to cream
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤2.5%

Description

It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text