Cyanuric fluoride, 98%, Thermo Scientific Chemicals
Cyanuric fluoride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyanuric fluoride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
100 g
Catalog number A15666.06
also known as A15666-06
Price (USD)/ Each
121.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
121.00
Add to cart
Cyanuric fluoride, 98%, Thermo Scientific Chemicals
Catalog numberA15666.06
Price (USD)/ Each
121.00
-
Add to cart
Chemical Identifiers
CAS675-14-9
IUPAC Nametrifluoro-1,3,5-triazine
Molecular FormulaC3F3N3
InChI KeyVMKJWLXVLHBJNK-UHFFFAOYSA-N
SMILESFC1=NC(F)=NC(F)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless or pale pink
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.3825-1.3865 @ 20°C (U.K. specification)
CommentPurchased in the U.K. and in other countries
Cyanuric fluoride acts as a fluorinating agent used in the conversion of carboxylic acids into acyl fluorides. It is used as a precursor for fibre-reactive dyes. It is a specific reagent for tyrosine residues in enzyme. Further, it is involved in the preparation of cyanuric acid by hydrolysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanuric fluoride acts as a fluorinating agent used in the conversion of carboxylic acids into acyl fluorides. It is used as a precursor for fibre-reactive dyes. It is a specific reagent for tyrosine residues in enzyme. Further, it is involved in the preparation of cyanuric acid by hydrolysis

Solubility
Miscible with chloroform, carbon tetrachloride,ether, dioxane and ketones.

Notes
May precipitate upon storage. Moisture sensitive. Incompatible strong oxidizing agents, strong bases and alcohols.
RUO – Research Use Only

General References:

  1. In combination with pyridine, converts carboxylic acids to acyl fluorides in high yields under mild conditions: Synthesis, 487 (1973). Similarly, protected (Fmoc) amino acids are converted in dichloromethane solution to amino acid fluorides. tert-Butyl ester and ether groups are also stable under these conditions. The resulting acyl fluorides are more stable to water than the corresponding acyl chlorides but are more reactive to amines, thus providing a useful peptide coupling method: J. Am. Chem. Soc., 112, 9651 (1990); Tetrahedron, 50, 5309 (1994); Appendix 6. For use of Fmoc amino acid fluorides in solid-phase peptide synthesis, see: Tetrahedron Lett., 32, 1303 (1991); J. Org. Chem., 59, 3275 (1994). For a review of peptide synthesis via amino acid halides, see: Acc. Chem. Res., 29, 268 (1996).
  2. Carboxylic acids can be converted to alcohols under mild conditions via the acyl fluoride and reduction with NaBH4 in the presence of MeOH. The method can be carried out in one pot and is applicable to N-protected amino acids and peptides for which other reduction methods are unsuitable: J. Org. Chem., 61, 6994 (1996).
  3. Reagent for deoxygenation of alkyl sulfoxides, cf Cyanuric chloride, L03442 : Synthesis, 221 (1980), and for dehydration, at room temperature in DMF, of Boc amino acid amides to nitriles: Tetrahedron. Lett., 38, 4221 (1997).
  4. Prabhaharan, M.; Prabakaran, A. R.; Gunasekaran, S.; Srinivasan, S. DFT studies on vibrational spectra, HOMO-LUMO, NBO and thermodynamic function analysis of cyanuric fluoride. Spectrochim. Acta, Part A 2015, 136, 494-503.
  5. Guo, W.; Pleixats, R.; Shafir, A.; Parella, T. Rhodium Nanoflowers Stabilized by a Nitrogen-Rich PEG-Tagged Substrate as Recyclable Catalyst for the Stereoselective Hydrosilylation of Internal Alkynes. Adv. Synth. Catal. 2015, 357 (1), 89-99.