Thermo Scientific Chemicals

Cyanuric fluoride, 98%, Thermo Scientific Chemicals

Catalog number: A15666.09
10 g, Each
Thermo Scientific Chemicals

Cyanuric fluoride, 98%, Thermo Scientific Chemicals

Catalog number: A15666.09
10 g, Each
Quantity
Catalog number: A15666.09
also known as A15666-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
675-14-9
IUPAC Name
trifluoro-1,3,5-triazine
Molecular Formula
C3F3N3
InChI Key
VMKJWLXVLHBJNK-UHFFFAOYSA-N
SMILES
FC1=NC(F)=NC(F)=N1
Appearance (Color)
Clear colorless or pale pink
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.3825-1.3865 @ 20°C (U.K. specification)
Comment
Purchased in the U.K. and in other countries

Description

Cyanuric fluoride acts as a fluorinating agent used in the conversion of carboxylic acids into acyl fluorides. It is used as a precursor for fibre-reactive dyes. It is a specific reagent for tyrosine residues in enzyme. Further, it is involved in the preparation of cyanuric acid by hydrolysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanuric fluoride acts as a fluorinating agent used in the conversion of carboxylic acids into acyl fluorides. It is used as a precursor for fibre-reactive dyes. It is a specific reagent for tyrosine residues in enzyme. Further, it is involved in the preparation of cyanuric acid by hydrolysis

Solubility
Miscible with chloroform, carbon tetrachloride,ether, dioxane and ketones.

Notes
May precipitate upon storage. Moisture sensitive. Incompatible strong oxidizing agents, strong bases and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text