Thermo Scientific Chemicals

D-(-)-2-Phenylglycine, 99%

Catalog number: A15669.14
25 g, Each
Thermo Scientific Chemicals

D-(-)-2-Phenylglycine, 99%

Catalog number: A15669.14
25 g, Each
Quantity
Catalog number: A15669.14
also known as A15669-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
875-74-1
IUPAC Name
(2R)-2-azaniumyl-2-phenylacetate
Molecular Formula
C8H9NO2
InChI Key
ZGUNAGUHMKGQNY-SSDOTTSWSA-N
SMILES
[NH3+][C@@H](C([O-])=O)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Optical Rotation
-154.5 ± 2.0? (C=1 in 1M HCl)

Description

D-(-)-2-Phenylglycine is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors. Used to prepare semisynthetic antibiotics amoxicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-2-Phenylglycine is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors. Used to prepare semisynthetic antibiotics amoxicillin.

Solubility
Soluble in water (0.3g/100mL) and aqueous acid.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text