Thermo Scientific Chemicals

Dicyclohexylamine, 98%, Thermo Scientific Chemicals

Catalog number: A15671.0F
2500 mL, Each
Thermo Scientific Chemicals

Dicyclohexylamine, 98%, Thermo Scientific Chemicals

Catalog number: A15671.0F
2500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15671.0F
also known as A15671-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101-83-7
IUPAC Name
N-cyclohexylcyclohexanamine
Molecular Formula
C12H23N
InChI Key
XBPCUCUWBYBCDP-UHFFFAOYSA-N
SMILES
C1CCC(CC1)NC1CCCCC1
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4825-1.4865 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%

Description

Dicyclohexylamine is used as an intermediate for the production of antioxidants and vulcanization accelerator. It is used as a catalyst for flexible polyurethane foams. It is also used in agrochemicals and textile chemicals. Used to constitute ionic liquid matrices for bacterial analysis in matrix assisted laser desorption/ionisation mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclohexylamine is used as an intermediate for the production of antioxidants and vulcanization accelerator. It is used as a catalyst for flexible polyurethane foams. It is also used in agrochemicals and textile chemicals. Used to constitute ionic liquid matrices for bacterial analysis in matrix assisted laser desorption/ionisation mass spectrometry.

Solubility
Sparingly soluble in water (0.8 g/L).

Notes
Air sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text