Thermo Scientific Chemicals

2-Hydroxy-3-methoxybenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A15672.22
100 g, Each
Thermo Scientific Chemicals

2-Hydroxy-3-methoxybenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A15672.22
100 g, Each
Quantity
Catalog number: A15672.22
also known as A15672-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
148-53-8
Form
Crystals or powder or crystalline powder
Appearance (Color)
Yellow to brown
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Melting Point (clear melt)
40-45?C

Description

o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.

Solubility
Solubility in methanol is almost transparent. Soluble in alcohol, methanol and dioxane (1g in 10 mL). Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text