1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).
Solubility
Soluble in water (partly) and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
RUO – Research Use Only
M F Lopez; W F Patton; W B Sawlivich; H Erdjument-Bromage; P Barry; K Gmyrek; T Hines; P Tempst; W M Skea. A glutathione S-transferase (GST) isozyme from broccoli with significant sequence homology to the mammalian theta-class of GSTs. Biochimica et Biophysica Acta. 1994, 1205 (1), 29-38 .
P Jemth; and B Mannervik. Kinetic characterization of recombinant human glutathione transferase T1-1, a polymorphic detoxication enzyme. Archives of Biochemistry and Biophysics. 1997, 348 (2), 247-254 .