Triethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals
Triethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15678.0B
Price (USD)/ Each
-
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS637-39-8
IUPAC Namehydrogen 2-[bis(2-hydroxyethyl)amino]ethan-1-ol chloride
Molecular FormulaC6H16ClNO3
InChI KeyHHLJUSLZGFYWKW-UHFFFAOYSA-N
SMILES[H+].[Cl-].OCCN(CCO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Assay (Titration ex Chloride)≥99.0 to ≤101.0%
Appearance (Color)White to cream or white to pale yellow
Identification (FTIR)Conforms
Protein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

General References:

  1. Russwurm, C.; Koesling, D.; Russwurm, M. Phosphodiesterase 10A Is Tethered to a Synaptic Signaling Complex in Striatum. J. Biol. Chem. 2015, 290 (19), 11936-11947.
  2. Opitz, S.; Hannika, F.; Krüger, T.; Rhode, H. The removal of Triton X-100 by dialysis is feasible!. Anal. Bioanal. Chem. 2015, 47 (2), 104-112.