Thermo Scientific Chemicals

p-Phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: A15680.30
250 g, Each
Thermo Scientific Chemicals

p-Phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: A15680.30
250 g, Each
Quantity
Catalog number: A15680.30
also known as A15680-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-50-3
IUPAC Name
benzene-1,4-diamine
Molecular Formula
C6H8N2
InChI Key
CBCKQZAAMUWICA-UHFFFAOYSA-N
SMILES
NC1=CC=C(N)C=C1
Form
Crystals or powder or crystalline powder or chunks or fused solid or flakes
Identification (FTIR)
Conforms
Appearance (Color)
Colorless to white to pale cream to cream to brown or pink to purple or pale gray to dark grey
Assay (GC)
≥96.0%
Melting Point (clear melt)
135.0-145.0?C

Description

p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

Solubility
Soluble in alcohol, chloroform, ether and hot benzene.

Notes
Store in cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text