Thermo Scientific Chemicals

Methyl anthranilate, 99%, Thermo Scientific Chemicals

Catalog number: A15681.36
500 g, Each
Thermo Scientific Chemicals

Methyl anthranilate, 99%, Thermo Scientific Chemicals

Catalog number: A15681.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15681.36
also known as A15681-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
134-20-3
IUPAC Name
methyl 2-aminobenzoate
Molecular Formula
C8H9NO2
InChI Key
VAMXMNNIEUEQDV-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1N
Assay (GC)
≥98.5%
Refractive Index
1.5815-1.5845 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow

Description

It is applied as a flavoring agent. Food companies use artificial MA in their candy and chewing gum to give them their grape flavor. Beverage companies have been adding it to the carbonated water and sugar in their soft drinks for the same reason.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text