Thermo Scientific Chemicals

Benzyl chloroformate, 95%, stab. with ca 0.1% sodium carbonate, Thermo Scientific Chemicals

Catalog number: A15682.18
50 g, Each
Thermo Scientific Chemicals

Benzyl chloroformate, 95%, stab. with ca 0.1% sodium carbonate, Thermo Scientific Chemicals

Catalog number: A15682.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15682.18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
501-53-1
IUPAC Name
benzyl carbonochloridate
Molecular Formula
C8H7ClO2
InChI Key
HSDAJNMJOMSNEV-UHFFFAOYSA-N
SMILES
ClC(=O)OCC1=CC=CC=C1
Refractive Index
1.5140-1.5200 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%

Description

Benzyl chloroformate is widely used as a reactive chemical intermediate in plastic, pharmaceutical, agricultural and organic chemicals. It is useful for the introduction of carboxybenzyl (cbz) protecting group for amines such as aniline in organic synthesis. It is also involved in the synthesis of 1,2,4-oxadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl chloroformate is widely used as a reactive chemical intermediate in plastic, pharmaceutical, agricultural and organic chemicals. It is useful for the introduction of carboxybenzyl (cbz) protecting group for amines such as aniline in organic synthesis. It is also involved in the synthesis of 1,2,4-oxadiazoles.

Solubility
Miscible with ether, acetone and benzene.

Notes
Store in cool place. Moisture sensitive.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text