Thermo Scientific Chemicals

Ethyl vinyl ether, 99%, stab., Thermo Scientific Chemicals

Catalog number: A15691.0F
2500 mL, Each
Thermo Scientific Chemicals

Ethyl vinyl ether, 99%, stab., Thermo Scientific Chemicals

Catalog number: A15691.0F
2500 mL, Each
Quantity
Catalog number: A15691.0F
also known as A15691-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-92-2
IUPAC Name
ethoxyethene
Molecular Formula
C4H8O
InChI Key
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
SMILES
CCOC=C
Appearance (Color)
Clear, colourless
Form
liquid
Assay (GC)
> 98.5%
Refractive Index
1.3755-1.3785

Description

Ethyl vinyl ether is used in fragrances, lubricating oil additives and spices preparation. It acts as an intermediate for sulfadiazine. It finds application in pharmaceuticals as anesthetics and analgesics. Further, it is used as a solvent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl vinyl ether is used in fragrances, lubricating oil additives and spices preparation. It acts as an intermediate for sulfadiazine. It finds application in pharmaceuticals as anesthetics and analgesics. Further, it is used as a solvent in organic synthesis.

Solubility
Miscible with common organic solvents. Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, oxygen and acids. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text