Thermo Scientific Chemicals

2-Bromo-1,3,5-triisopropylbenzene, 96%, Thermo Scientific Chemicals

Catalog number: A15692.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-1,3,5-triisopropylbenzene, 96%, Thermo Scientific Chemicals

Catalog number: A15692.14
25 g, Each
Quantity
Catalog number: A15692.14
also known as A15692-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21524-34-5
IUPAC Name
2-bromo-1,3,5-tris(propan-2-yl)benzene
Molecular Formula
C15H23Br
InChI Key
FUMMYHVKFAHQST-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(C(C)C)=C(Br)C(=C1)C(C)C
Identification (FTIR)
Conforms
Refractive Index
1.5195-1.5255 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥95.0%

Description

2-Bromo-1,3,5-triisopropylbenzene is used in Suzuki reaction. It is found to effect highly selective ring closures of homoallylic alcohols to substituted tetrahydrofurans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,3,5-triisopropylbenzene is used in Suzuki reaction. It is found to effect highly selective ring closures of homoallylic alcohols to substituted tetrahydrofurans.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text