Methyl o-toluate, 99%, Thermo Scientific Chemicals
Methyl o-toluate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl o-toluate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A15695.36
also known as A15695-36
Price (USD)/ Each
459.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
459.00
Add to cart
Methyl o-toluate, 99%, Thermo Scientific Chemicals
Catalog numberA15695.36
Price (USD)/ Each
459.00
-
Add to cart
Chemical Identifiers
CAS89-71-4
IUPAC Namemethyl 2-methylbenzoate
Molecular FormulaC9H10O2
InChI KeyWVWZECQNFWFVFW-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.5175-1.5205 @ 20°C
Assay (GC)≥98.5%
It has been used in the preparation of 2-(azidomethyl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used in the preparation of 2-(azidomethyl)benzoic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
RUO – Research Use Only

General References:

  1. Wada T, et al. 2-(Azidomethyl) benzoyl as a new protecting group in nucleosides.Tetrahedron Lett.,2001,42(6), 1069-72.
  2. Fache F, et al. A catalytic stereo-and chemo-selective method for the reduction of substituted aromatics.Tetrahedron Lett.,1995,36(6), 885-88.