Thermo Scientific Chemicals

4-Methoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A15697.0B
1000 g, Each
Thermo Scientific Chemicals

4-Methoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A15697.0B
1000 g, Each
Quantity
Catalog number: A15697.0B
also known as A15697-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-09-4
Appearance (Color)
White to pale cream
Form
Powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
181-191?C
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

4-Methoxybenzoic acid is used in oxidation and reduction of cytochrome c in solution through different self-assembled monolayers on gold electrodes using cyclic voltammetry. p-Anisic acid has antiseptic properties. It is also used as an intermediate in the preparation of more complex organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzoic acid is used in oxidation and reduction of cytochrome c in solution through different self-assembled monolayers on gold electrodes using cyclic voltammetry. p-Anisic acid has antiseptic properties. It is also used as an intermediate in the preparation of more complex organic compounds.

Solubility
Soluble in water at 20°C 0.3g/L. Soluble in alcohol, ethyl acetate and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text