9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.
Solubility
Soluble in water (0,0003 mg/L at 25°C) and toluene. Slightly soluble in benzene.
Notes
Mechanical exhaust required. When not in use, tightly seal the container and store in a dry, cool place.
RUO – Research Use Only
K Takegoshi; S Nakamura. Solid-state photodimerization of 9-methylanthracene as studied by solid-state 13C NMR. Solid State Nuclear Magnetic Resonance. 1998, 11 (3-4), 189-196.
B V Pepich; J B Callis; D H Burns; M Gouterman; D A Kalman. Capillary gas chromatography/pulsed supersonic jet/fluorescence excitation spectroscopy for the identification of methylanthracenes in a complex environmental sample. Analytical chemistry. 1998, 58 (13), 2825-2830.