Thermo Scientific Chemicals

2-Butyn-1-ol, 98%, Thermo Scientific Chemicals

Catalog number: A15709.22
100 g, Each
Thermo Scientific Chemicals

2-Butyn-1-ol, 98%, Thermo Scientific Chemicals

Catalog number: A15709.22
100 g, Each
Quantity
Catalog number: A15709.22
also known as A15709-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
764-01-2
IUPAC Name
but-2-yn-1-ol
Molecular Formula
C4H6O
InChI Key
NEEDEQSZOUAJMU-UHFFFAOYSA-N
SMILES
CC#CCO
Refractive Index
1.4510-1.4550 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Form
Liquid
Identification (FTIR)
Conforms

Description

2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

Solubility
Miscible with chloroform and methanol.

Notes
Incompatible with oxidizing agents, acids, acid anhydrides and copper.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text