Thermo Scientific Chemicals

(+/-)-1-Phenylethylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A15711.AP
500 mL, Each
Thermo Scientific Chemicals

(+/-)-1-Phenylethylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A15711.AP
500 mL, Each
Quantity
Catalog number: A15711.AP
also known as A15711-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
618-36-0
IUPAC Name
1-phenylethan-1-amine
Molecular Formula
C8H11N
InChI Key
RQEUFEKYXDPUSK-UHFFFAOYNA-N
SMILES
CC(N)C1=CC=CC=C1
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%
Refractive Index
1.5245-1.5285 @ 20?C

Description

Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.

Solubility
Soluble in water at 20°C 4.2 g/L.

Notes
Air sensitive. Store away from air, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text