Thermo Scientific Chemicals

Dextrin, precipitated by alcohol, Thermo Scientific Chemicals

Catalog number: A15717.22
100 g, Each
Thermo Scientific Chemicals

Dextrin, precipitated by alcohol, Thermo Scientific Chemicals

Catalog number: A15717.22
100 g, Each
Quantity
Catalog number: A15717.22
also known as A15717-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-53-9
IUPAC Name
(3R,4S,5S,6R)-2-{[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C18H32O16
InChI Key
FYGDTMLNYKFZSV-MRCIVHHJSA-N
SMILES
OC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](O)C(O[C@H]3[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Appearance (Color)
White to yellow to brown
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Loss on Drying
<11.0% (105°C/constant wt.)
Sulfated Ash
≤1.0%

Description

Dextrin is used as adhesive in the manufacture of gummed tapes, textiles and paper. It is used in producing nutritional products. It is also used as a absorbents, binding agents and viscosity controlling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text