Thermo Scientific Chemicals

2-Amino-5-chlorobenzophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A15720.0E
2500 g, Each
Thermo Scientific Chemicals

2-Amino-5-chlorobenzophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A15720.0E
2500 g, Each
Quantity
Catalog number: A15720.0E
also known as A15720-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
719-59-5
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥98.0%
Melting Point (clear melt)
95.0-102.0?C
Identification (FTIR)
Conforms

Description

It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.

Solubility
Solubility in methanol gives very faint turbidity. Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text