Thermo Scientific Chemicals

Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals

Catalog number: A15722.06
5 g, Each
Thermo Scientific Chemicals

Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals

Catalog number: A15722.06
5 g, Each
Quantity
Catalog number: A15722.06
also known as A15722-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
476-66-4
IUPAC Name
6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
Molecular Formula
C14H6O8
InChI Key
AFSDNFLWKVMVRB-UHFFFAOYSA-N
SMILES
OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O
Appearance (Color)
White to cream to yellow to green to brown to grey
Comment
May contain up to 12% water.
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤12.0%
Form
Powder

Description

Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

Solubility
Slightly soluble in water. Soluble in 1M sodium hydroxide (10 mg/ml), EtOH, and pyridine.

Notes
Air sensitive and light sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container. Store in dark and away form air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text