Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals
Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A15722.22
also known as A15722-22
Price (USD)/ Each
496.65
Online exclusive
552.00 
Save 55.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
496.65
Online exclusive
552.00 
Save 55.35 (10%)
Add to cart
Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals
Catalog numberA15722.22
Price (USD)/ Each
496.65
Online exclusive
552.00 
Save 55.35 (10%)
-
Add to cart
Chemical Identifiers
CAS476-66-4
IUPAC Name6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
Molecular FormulaC14H6O8
InChI KeyAFSDNFLWKVMVRB-UHFFFAOYSA-N
SMILESOC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to green to brown to grey
CommentMay contain up to 12% water.
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤12.0%
FormPowder
View more
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

Solubility
Slightly soluble in water. Soluble in 1M sodium hydroxide (10 mg/ml), EtOH, and pyridine.

Notes
Air sensitive and light sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container. Store in dark and away form air.
RUO – Research Use Only

General References:

  1. Patrice Njomnang Soh: Benoît Witkowski; David Olagnier; Marie-Laure Nicolau; Maria-Concepcion Garcia-Alvarez; Antoine Berry; and Françoise Benoit-Vical. In Vitro and In Vivo Properties of Ellagic Acid in Malaria Treatment. Antimicrob. Agents Chemother. 2009, 31100-1106.
  2. Sungwoo Kim; Satoru K. Nishimoto; Joel D. Bumgardnerd; Warren O. Haggardd; M. Waleed Gabere; Yunzhi Yang. A chitosan/β-glycerophosphate thermo-sensitive gel for the delivery of ellagic acid for the treatment of brain cancer. Biomaterials. 2010, 31 (14), 4157-4166.