Thermo Scientific Chemicals

Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.), Thermo Scientific Chemicals

Catalog number: A15731.36
500 g, Each
Thermo Scientific Chemicals

Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.), Thermo Scientific Chemicals

Catalog number: A15731.36
500 g, Each
Quantity
Catalog number: A15731.36
also known as A15731-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149-44-0
IUPAC Name
sodium hydroxymethanesulfinate
Molecular Formula
CH3NaO3S
InChI Key
XWGJFPHUCFXLBL-UHFFFAOYSA-M
SMILES
[Na+].OCS([O-])=O
Sulfated Ash
> 50.0%
Form
powder
Comment
Alkalinity as NaOH < 2.0%
Appearance (Color)
White
Assay (ex Sulphated Ash)
> 83%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text