Thermo Scientific Chemicals

4-Nitrobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A15742.14
25 g, Each
Thermo Scientific Chemicals

4-Nitrobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A15742.14
25 g, Each
Quantity
Catalog number: A15742.14
also known as A15742-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
619-73-8
IUPAC Name
(4-nitrophenyl)methanol
Molecular Formula
C7H7NO3
InChI Key
JKTYGPATCNUWKN-UHFFFAOYSA-N
SMILES
OCC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
Pale cream to cream to yellow
Form
Powder or lumps
Assay (GC)
≥98.5%
Melting Point (clear melt)
90.0-97.0?C

Description

4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.

Solubility
Soluble in water (2 mg/ml at 20°C).

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text