Thermo Scientific Chemicals

2-Chloroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A15758.0C
10,000 g, Each
Thermo Scientific Chemicals

2-Chloroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A15758.0C
10,000 g, Each
Quantity
Catalog number: A15758.0C
also known as A15758-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-51-2
IUPAC Name
2-chloroaniline
Molecular Formula
C6H6ClN
InChI Key
AKCRQHGQIJBRMN-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1Cl
Appearance (Color)
Clear colorless to orange or brown
Refractive Index
1.5860-1.5900 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%

Description

2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

Solubility
Miscible with alcohol, ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text