6-Bromohexanoic acid, 98+%, Thermo Scientific Chemicals
6-Bromohexanoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromohexanoic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A15761.22
also known as A15761-22
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
Add to cart
6-Bromohexanoic acid, 98+%, Thermo Scientific Chemicals
Catalog numberA15761.22
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS4224-70-8
IUPAC Name6-bromohexanoate
Molecular FormulaC6H10BrO2
InChI KeyNVRVNSHHLPQGCU-UHFFFAOYSA-M
SMILES[O-]C(=O)CCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or orange
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)30.0-36.0?C
Assay (Silylated GC)≥98.0%
6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.

Solubility
Soluble in methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Topel, S. D.; Topel, O.; Bostancıoglu, R. B.; Koparal, A. T. Synthesis and characterization of Bodipy functionalized magnetic iron oxide nanoparticles for potential bioimaging applications. Colloids Surf., B 2015, 128, 245-253.
  2. Snelling, R.; Saavedra, J. Z.; Bayrasy, P.; Abdollahian, Y.; Singaram, B. Binary reducing agents containing dichloroindium hydride for the selective, partial, or tandem reductions of bifunctional compounds consisting of halo-nitriles, halo-esters and halo-carboxylic acids. Org. Chem. Front. 2015, 2 (2), 133-140.