Thermo Scientific Chemicals

4-Cyanobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A15764.06
5 g, Each
Thermo Scientific Chemicals

4-Cyanobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A15764.06
5 g, Each
Quantity
Catalog number: A15764.06
also known as A15764-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1503-49-7
IUPAC Name
4-benzoylbenzonitrile
Molecular Formula
C14H9NO
InChI Key
YSZWJJANSNFQMM-UHFFFAOYSA-N
SMILES
O=C(C1=CC=CC=C1)C1=CC=C(C=C1)C#N
Form
Powder
Melting Point (clear melt)
108.0-115.0?C
Assay (GC)
≥97.5%
Appearance (Color)
White to cream to pale yellow

Description

Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)quadricyclane(Q) and QN isomerization. It is employed as a triplet sensitizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)→quadricyclane(Q) and Q→N isomerization. It is employed as a triplet sensitizers.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text