Thermo Scientific Chemicals

trans-Crotonic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15765.36
500 g, Each
Thermo Scientific Chemicals

trans-Crotonic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15765.36
500 g, Each
Quantity
Catalog number: A15765.36
also known as A15765-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-93-7
IUPAC Name
(2E)-but-2-enoic acid
Molecular Formula
C4H6O2
InChI Key
LDHQCZJRKDOVOX-NSCUHMNNSA-N
SMILES
C\C=C\C(O)=O
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder or crystalline flakes or pellets
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms

Description

trans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.

Solubility
Soluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text